Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135927.1 | NP_001129399.1 | 1 | 815 | 95 | |
XM_012179364.3 | XP_012034754.2 | 2 | 903 | 95 | ||
XM_012179365.3 | XP_012034755.2 | 2 | 957 | 95 | ||
XM_015096245.2 | XP_014951731.2 | 2 | 1066 | 95 | ||
GenBank mRNA | AM981304.1 | - | 1 | 815 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |