Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009432.1 | NP_001009432.1 | 5 - 6 | 714 | 79 | |
GenBank mRNA | U68486.1 | - | 5 - 6 | 714 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |