Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YEATS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163059.1 | NP_001156531.1 | 2 - 3 | 170 | 113 | |
GenBank mRNA | FJ958351.1 | - | 2 - 3 | 170 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |