Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009796.1 | NP_001009796.1 | 6 - 7 | 1179 | 82 | |
XM_027975567.1 | XP_027831368.1 | 5 - 6 | 1136 | 82 | ||
GenBank mRNA | X03883.1 | - | 1179 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |