Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126364.1 | NP_001119836.1 | 1 - 2 | 55 | 74 | |
XM_027957540.1 | XP_027813341.1 | 2 - 3 | 311 | 74 | ||
XM_027957541.1 | XP_027813342.1 | 2 - 3 | 1559 | 74 | ||
XM_027957542.1 | XP_027813343.1 | 2 - 3 | 150 | 74 | ||
XM_027957543.1 | XP_027813344.1 | 2 - 3 | 157 | 74 | ||
XM_027957544.1 | XP_027813345.1 | 2 - 3 | 322 | 74 | ||
GenBank mRNA | EU596593.1 | - | 1 - 2 | 55 | 74 |