Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040289.1 | NP_001035379.1 | 3 - 4 | 293 | 68 | |
GenBank mRNA | DQ458790.1 | - | 3 - 4 | 293 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |