Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287469.1 | NP_001274398.1 | 2 - 3 | 133 | 89 | |
XM_027965740.1 | XP_027821541.1 | 1 - 2 | 358 | 89 | ||
GenBank mRNA | KF408269.1 | - | 2 - 3 | 133 | 89 |