Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011704.4 | XP_004011753.2 | 5 - 6 | 811 | 72 | |
XM_012183770.2 | XP_012039160.2 | 3 - 4 | 277 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |