Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961613.1 | XP_027817414.1 | 4 - 5 | 494 | 97 | |
XM_027961614.1 | XP_027817415.1 | 3 - 4 | 1011 | 97 | ||
XM_027961615.1 | XP_027817416.1 | 3 - 4 | 463 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |