Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101239.2 | XP_014956725.2 | 4 - 5 | 1560 | 83 | |
XM_027979991.1 | XP_027835792.1 | 3 - 4 | 840 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |