Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973546.1 | XP_027829347.1 | 5 - 6 | 730 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |