Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973546.1 | XP_027829347.1 | 8 - 9 | 1134 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |