Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957271.1 | XP_027813072.1 | 4 - 5 | 660 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |