Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970127.1 | XP_027825928.1 | 2 - 3 | 305 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |