Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100555.3 | XP_011955945.2 | 1 - 2 | 378 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |