Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956338.1 | XP_027812139.1 | 3 - 4 | 728 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |