Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004503.4 | XP_004004552.1 | 3 - 4 | 469 | 102 | |
XM_004004505.4 | XP_004004554.1 | 3 - 4 | 396 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |