Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003657.4 | XP_004003706.1 | 3 - 4 | 409 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |