Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3RF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966794.1 | XP_027822595.1 | 9 - 10 | 3376 | 89 |