Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096738.2 | XP_014952224.2 | 3 - 4 | 664 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |