Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972368.1 | XP_027828169.1 | 3 - 4 | 281 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |