Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005705.4 | XP_004005754.1 | 1 - 2 | 303 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Ovis aries |