Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009470.4 | XP_004009519.2 | 4 - 5 | 499 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |