Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095281.2 | XP_011950671.2 | 13 - 14 | 1718 | 96 | |
XM_012095282.3 | XP_011950672.2 | 13 - 14 | 1682 | 96 | ||
XM_012095283.2 | XP_011950673.2 | 11 - 12 | 1344 | 96 | ||
XM_012095284.2 | XP_011950674.2 | 10 - 11 | 1272 | 96 |