Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008484.4 | XP_004008533.1 | 4 - 5 | 564 | 65 |
Gene Symbol: LOC114114906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970479.1 | XP_027826280.1 | 1 | 558 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |