Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012154.4 | XP_004012203.3 | 4 - 5 | 777 | 101 | |
XM_015098160.2 | XP_014953646.1 | 4 - 5 | 811 | 101 | ||
XM_015098161.2 | XP_014953647.1 | 4 - 5 | 895 | 101 | ||
XM_015098162.2 | XP_014953648.1 | 4 - 5 | 749 | 101 | ||
XM_015098163.2 | XP_014953649.1 | 5 - 6 | 915 | 101 | ||
XM_015098164.2 | XP_014953650.1 | 5 - 6 | 854 | 101 | ||
XM_027973640.1 | XP_027829441.1 | 5 - 6 | 950 | 101 | ||
XM_027973641.1 | XP_027829442.1 | 5 - 6 | 887 | 101 | ||
XM_027973643.1 | XP_027829444.1 | 5 - 6 | 826 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |