Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972609.1 | XP_027828410.1 | 3 - 4 | 422 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |