Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102701.2 | XP_014958187.2 | 4 - 5 | 689 | 67 | |
XM_015102702.2 | XP_014958188.2 | 4 - 5 | 693 | 67 | ||
XM_015102703.2 | XP_014958189.2 | 4 - 5 | 624 | 67 | ||
XM_015102704.2 | XP_014958190.2 | 4 - 5 | 550 | 67 | ||
XM_027958635.1 | XP_027814436.1 | 4 - 5 | 684 | 67 | ||
GenBank mRNA | AF184163.1 | - | 2 - 3 | 270 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |