Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975409.1 | XP_027831210.1 | 3 - 4 | 1021 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |