Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976124.1 | XP_027831925.1 | 7 - 8 | 908 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |