Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011087.3 | XP_004011136.2 | 5 - 6 | 792 | 91 | |
XM_027972084.1 | XP_027827885.1 | 5 - 6 | 762 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |