Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969302.1 | XP_027825103.1 | 4 - 5 | 1484 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |