Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979963.1 | XP_027835764.1 | 5 - 6 | 851 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |