Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004232.4 | XP_004004281.1 | 3 - 4 | 415 | 103 | |
XM_012120695.3 | XP_011976085.1 | 3 - 4 | 405 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |