Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104546.3 | XP_011959936.2 | 2 - 3 | 1788 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |