Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004342.4 | XP_004004391.2 | 2 - 3 | 530 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |