Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPSCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098906.2 | XP_014954392.2 | 8 - 9 | 1135 | 56 | |
XM_027974287.1 | XP_027830088.1 | 8 - 9 | 1343 | 56 | ||
XM_027974288.1 | XP_027830089.1 | 8 - 9 | 1105 | 56 | ||
XM_027974289.1 | XP_027830090.1 | 7 - 8 | 1058 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |