Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965894.1 | XP_027821695.1 | 5 - 6 | 535 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |