Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185208.2 | XP_012040598.1 | 2 - 3 | 350 | 65 | |
XM_015098465.2 | XP_014953951.1 | 2 - 3 | 472 | 65 | ||
XM_015098466.2 | XP_014953952.1 | 2 - 3 | 472 | 65 | ||
XM_027975342.1 | XP_027831143.1 | 2 - 3 | 332 | 65 | ||
XM_027975343.1 | XP_027831144.1 | 2 - 3 | 544 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |