Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102307.2 | XP_014957793.1 | 5 - 6 | 739 | 85 | |
XM_027958096.1 | XP_027813897.1 | 4 - 5 | 1122 | 85 | ||
XM_027958097.1 | XP_027813898.1 | 5 - 6 | 739 | 85 | ||
XM_027958098.1 | XP_027813899.1 | 5 - 6 | 736 | 85 | ||
XM_027958099.1 | XP_027813900.1 | 5 - 6 | 736 | 85 | ||
XM_027958100.1 | XP_027813901.1 | 5 - 6 | 942 | 85 | ||
XM_027958101.1 | XP_027813902.1 | 5 - 6 | 736 | 85 | ||
XM_027958102.1 | XP_027813903.1 | 5 - 6 | 742 | 85 | ||
XM_027958103.1 | XP_027813904.1 | 5 - 6 | 740 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |