Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012127127.2 | XP_011982517.1 | 1 | 963 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |