Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003498.4 | XP_004003547.3 | 1 | 185 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |