Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956849.1 | XP_027812650.1 | 3 - 4 | 444 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |