Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101116620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103601.2 | XP_014959087.2 | 2 - 3 | 595 | 71 | |
XM_027961070.1 | XP_027816871.1 | 2 - 3 | 595 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |