Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015021.4 | XP_004015070.1 | 1 - 2 | 271 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |