Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022164.4 | XP_004022213.1 | 2 - 3 | 765 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |