Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968889.1 | XP_027824690.1 | 1 | 793 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |