Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969311.1 | XP_027825112.1 | 2 - 3 | 701 | 75 | |
XM_027969322.1 | XP_027825123.1 | 2 - 3 | 693 | 75 | ||
XM_027969328.1 | XP_027825129.1 | 2 - 3 | 706 | 75 | ||
XM_027969345.1 | XP_027825146.1 | 2 - 3 | 698 | 75 | ||
XM_027969351.1 | XP_027825152.1 | 2 - 3 | 702 | 75 |