Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973995.1 | XP_027829796.1 | 5 - 6 | 817 | 83 | |
XM_027973996.1 | XP_027829797.1 | 3 - 4 | 1488 | 83 | ||
XM_027973997.1 | XP_027829798.1 | 5 - 6 | 541 | 83 |