Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976392.1 | XP_027832193.1 | 2 - 3 | 543 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |